missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MERS-CoV Spike RBD Antibody (AT18D10) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26835JF549
This item is not returnable.
View return policy
Description
MERS-CoV Spike RBD Monoclonal antibody specifically detects MERS-CoV Spike RBD in MERS-CoV samples. It is validated for ELISA,Western Blot
Specifications
| MERS-CoV Spike RBD | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| MERS-CoV | |
| Purified |
| ELISA, Western Blot | |
| AT18D10 | |
| 50 mM Sodium Borate | |
| Recombinant MERS-CoV Spike RBD (Uniprot#: K0BRG7, Accession#: AFS88936, 358-606aa) purified from Baculovirus | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction