missing translation for 'onlineSavingsMsg'
Learn More

Tiadinil, Honeywell Fluka™

Product Code. 15971081 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY

Specifications

CAS 223580-51-6
Packaging Glass Bottle
Quantity 25 mg
For Use With (Application) Fungicides and Bactericides
Physical Form Crystalline Powder
Grade Analytical Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.