missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MEN1/CEN11p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
1515.00€ - 2495.00€
Specifications
Content And Storage | Store at 4°C in the dark. |
---|---|
Label or Dye | FITC, Texas Red |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections) |
Includes | DAPI Counterstain (1500 ng/mL ) 250 uL |
Location | 11q13, 11p11.12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16063045
|
Abnova™
FG0040.200uL |
200 μL |
2495.00€
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16053045
|
Abnova™
FG0040.100uL |
100 μL |
1515.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifications
Store at 4°C in the dark. | |
Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections) | |
11q13, 11p11.12 | |
MEN1/CEN11p FISH Probe |
FITC, Texas Red | |
DAPI Counterstain (1500 ng/mL ) 250 uL | |
MEN1, CEN11p |