Learn More
MED20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17598-1-AP-20UL
 View more versions of this product
Product Code. 16837534
  • 145.00€ / 20µL
Description
MED20 is a subunit of the Mediator complex, a multiprotein coactivator of RNA transcription that interacts with DNA-bound transcriptional activators, RNA polymerase II, and general initiation factors.
Specifications
Specifications
MED20
Polyclonal
Unconjugated
MED20
hTRFP, MED20, mediator complex subunit 20, PRO0213, TRF proximal protein homolog, TRFP
Rabbit
Antigen Affinity Chromatography
RUO
316209, 56771, 9477
-20°C
Liquid
Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5XIE9, Q9H944, Q9R0X0
Med20
MED20 Fusion Protein Ag11598
20 μL
Primary
Mouse, Human, Rat