Learn More
Buerkle™ Measuring Jug Industrial with Closed Handle
Light and handy polypropylene measuring jug is suitable for industrial applications.
6.50€ - 21.90€
Specifications
Material Polypropylene
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
16696553
View Documents
Buerkle™
7204-1000
1000 mL
6.50€
Each
In Stock
to see stock.
Add to basket
16606563
View Documents
Buerkle™
7204-5000
5000 mL
21.90€
Each
In Stock
to see stock.
Add to basket
Description
Description
The measuring jug has two measuring scales, embossed transparent and with blue lettering. With ergonomic handle, available in closed design for special stability or with open handle for stacking to save space.The measuring jug is made of polypropylene and is therefore highly resistant to acids, bases and chemicals.
  • PP, transparent
  • With embossed graduation printed in blue
Specifications
Specifications
Polypropylene