Learn More
WTW™ 50mL Measuring Beaker
For respirometric measurements of BSB, soil respiration, degradability tests and biogas examinations.
Brand:  WTW™ 108064
Product Code. 10736642
  • 13.50€ / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Tested for respective applications
  • Supports accurate measurements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50mL
Measuring Cup/Beaker
Respirometric measurements, soil respiration, degradability tests, biogas examinations