missing translation for 'onlineSavingsMsg'
Learn More
Learn More
McFarland Standard Set, Pro-Lab Diagnostics™
Click to view available options
Quantity:
1 Set
Unit Size:
Each
Specifications
Specifications
| Physical Form | Liquid |
| Color | Opaque |
| Solubility | Soluble in water |
| Quantity | 1 Set |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction