Learn More
MBL™ Borosilicate Glass Class A Col Lisse
Product Code. 12124432
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
2000 mL
5 L
Height (Metric):
70 mm
90 mm
110 mm
140 mm
170 mm
210 mm
220 mm
260 mm
370 mm
475 mm
Tolerance:
±0.025 mL
±0.04 mL
±0.1 mL
±0.15 mL
±0.25 mL
±0.6 mL
±1.2 mL
Unit Size:
Each
Pack of 2
Pack of 5
Product Code. 12124432
Brand: MBL FRD222
Only null left
Add to basket
Add to basket
Calibrated to Class A tolerances. MBL™ Borosilicate Glass Class A Col Lisse is iIndividually laser etched with serial number, batch code and date of manufacture
  • Complies with ISO 1042 and DIN 12664
  • Finely calibrated to Class A tolerances
  • Each flask is laser etched with an individual serial number, batch code and date of manufacture
  • Manufactured from chemically resistant borosilicate glass
  • Conformity 'H' certified
  • Inscriptions in blue enamel
  • Supplied with polyethylene stopper
  • Batch certificate available to download online
  • Complies with ISO 1042
  • With plain neck to accept rubber stopper
  • Manufactured from borosilicate glass
  • Finely calibrated to Class A tolerances
  • Conformity 'H' certified
  • Individually laser etched with serial number, batch code and date of manufacture
  • Batch certificate available to download online
  • Blue enamel inscriptions
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Class A, ISO 1042, Conformity 'H' certified
Closure Material Rubber
Capacity (Metric) 5 L
Material Borosilicate Glass
Height (Metric) 475 mm
Neck Style Plain
Tolerance ±1.2 mL
Graduated Yes
Class A