Learn More
Ohaus™ MB120 Moisture Analyzer
Measures moisture content with increased precision and efficiency.
Brand:  Ohaus™ 30241169
3808.30 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Product Code. 15535029
  • 5180.00€ / 1 unit
Explore more special offers
Description
Description
  • Easily analyze specific material samples and determine the optimum drying temperature
  • 100 methods library makes testing faster
  • Store up to 1000 results for all methods for statistical analysis
  • Set up and lock temperature settings and methods parameters with admin access
  • Features wide, bright touchscreen display and intuitive menu navigation for fast, accurate moisture analysis
  • Fast infrared halogen heating technology
  • Uniform drying for precise, repeatable results
  • Flexible and powerful
  • Four drying profiles to ensure accuracy: Fast, Standard, Ramp, Step
  • Shut-Off criteria for automatic test completion
  • Features pan handler and removable parts for convenient maintenance
Certifications
CE, UL listed, CSA approved and FCC approved
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Ohaus™ MB120 Moisture Analyzer > MB120 Moisture Analyzer