Learn More
Thermo Scientific™ Maxima H Minus Reverse Transcriptase Promotion
A highly thermostable, high processivity reverse transcriptase lacking RNase H activity
Brand:  Thermo Scientific™ EP0752
 View more versions of this product
Product Code. 15777496
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Thermostable - 90% active after incubation at 50°C for 60 minutes in a reaction mixture.
  • Active up to 65°C
  • High yields of full-length cDNA up to 20 kb
  • High sensitivity - reproducible cDNA synthesis from a wide range of starting total RNA amounts (1 pg to 5 μg)
  • Efficient - complete cDNA synthesis in 15 to 30 minutes
  • Increased resistance to common reaction inhibitors
  • Incorporates modified nucleotides
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250 mM Tris-HCl buffer (pH 8.3 at 25°C), 37 5mM KCl, 15 mM MgCl2, 50 mM DTT
10,000 U
5X
-20°C
cDNA Synthesis, RT-PCR, Gene Expression