Learn More
3M™ Maxim™ DX Ballistic Safety Glasses
Click to view available options
Lens Tint:
Amber
Bronze
Clear
Unit Size:
Pack of 20
Description
- Safety spectacles that exceed the Military VO test regimen for high velocity impact
- DX coating provides enhanced anti-scratch and anti-fog protection
- Spectacles with ventilation channel across the lens to minimize misting
- Soft, adjustable nose bridge and a soft-touch brow guard
- Adjustable temple lengths and pantoscopic (angle) adjustments
- Design provides excellent coverage and good field of vision
- Low-profile design increases compatibility with helmets and hearing protection
Specifications
Specifications
| Certifications/Compliance | EN 166:2001 |
| Lens Type | DX |
| Lens Tint | Amber |
| Product Type | Ballistic Safety Glasses |
| For Use With (Application) | Construction |
| Frame Color | Black, Gray |
| Lens Material | Polycarbonate |