Learn More
3M™ Maxim™ DX Ballistic Safety Glasses
Product Code. 15799657
Click to view available options
Lens Tint:
Amber
Bronze
Clear
Unit Size:
Pack of 20
Product Code. 15799657
Brand: 3M™ 7000030023
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
3M™ Maxim™ DX Ballistic Safety Glasses are designed specifically to wear in combination with a helmet and or ear muffs.
  • Safety spectacles that exceed the Military VO test regimen for high velocity impact
  • DX coating provides enhanced anti-scratch and anti-fog protection
  • Spectacles with ventilation channel across the lens to minimize misting
  • Soft, adjustable nose bridge and a soft-touch brow guard
  • Adjustable temple lengths and pantoscopic (angle) adjustments
  • Design provides excellent coverage and good field of vision
  • Low-profile design increases compatibility with helmets and hearing protection
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166:2001
Lens Type DX
Lens Tint Amber
Product Type Ballistic Safety Glasses
For Use With (Application) Construction
Frame Color Black, Gray
Lens Material Polycarbonate