missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Mastercycler Nexus GX2, EU-Plug
PCR cycler with the capacity to run two simultaneous and independent protocols.
Brand: Eppendorf™ 6336000015
This item is not returnable.
View return policy
Description
- Large block for large assays - small block for small assays
- Intuitive graphic programming
- Universal block for strips, 0.1 mL, 0.2 mL, and 0.5 mL PCR tubes
- Small footprint
- Optional gradient for PCR optimization
- E-mail notification
- flexlid™ concept allows use of all types of consumables with automatic height adjustment of the lid
Specifications
37 to 110°C (Lid), 4 to 99°C (Block) | |
25 × 41.2 × 33 cm | |
230 V, 50/60 Hz | |
CAN_in/CAN_out, Ethernet, USB | |
Yes |
EU Plug | |
64/32 × 0.2 mL PCR Tubes or 24/10 × 0.5 mL PCR Tube Positions | |
Standard PCR, Optimizing a PCR | |
>700 | |
10.5 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction