Learn More
Carver™ Manual Pellet Press
Carver™ Pellet Presses compact homogeneous powder into a suitable pellet sample.
Brand:  Carver™ 4555CE
Product Code. 11999996
  • 34880.00€ / Each
Explore more special offers
Special packaging extra freight cost. Please contact us for quotation Récupel-bijdrage uitgesloten
Description
Description
  • Evacuation port for connection to vacuum source
  • Oversize pressure gauge for ease of viewing
  • Inegral safety shields
Specifications
Specifications
Pellet Press
Sample Prep
31 mm die
40 US tons
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Carver™ Manual Pellet Press > Pellet Press; Normal Force: 40 US tons