missing translation for 'onlineSavingsMsg'
Learn More

Maleate Standard for Ion Chromatography, 1000 mg/L in water, Honeywell Fluka™

Product Code. 16604234 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 16604234

Brand: Honeywell Fluka E3026100ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Maleate Standard for Ion Chromatography
Density 1.01 g/cm3
Quantity 100 mL
Grade Standards for Ion Chromatography
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.