Learn More
Rogo Sampaic™ Malassez Counting Cells
For determining the number of particles per volume unit of a liquid. Rogo Sampaic™ Malassez Counting Cells have a simple or double grid with a depth of 0.2mm.
Brand:  Rogo Sampaic™ ZMB024
 View more versions of this product
Product Code. 11740835
  • 128.00€ / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For blood samples
  • Provides for accurate manual measurements
Specifications
Specifications
Malassez Cell
1Each
0.2mm
Simple
Blood
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers