Learn More
Invitrogen™ MagniSort™ Streptavidin Negative Selection Beads
Designed for the magnetic separation of cells by negative selection. Undesired cells are bound by a cocktail of biotinylated antibodies and then streptavidin coated magnetic beads.
Brand:  Invitrogen™ MSNB-6002-71
 View more versions of this product
Product Code. 15562567
  • 95.75€ / 25 tests
In Stock
to see stock.
Explore more special offers
Add to basket
Alert:
Contains Sodium azide
Description
Description
  • Clone and concentration of biotinylated antibody for undesired cell labeling, as well as concentration of MagniSort™ Streptavidin Negative Selection Beads must be determined empirically
  • Please refer to the protocol for details on how to optimize both the antibody or antibody cocktail and MagniSort™ Streptavidin Negative Selection Beads.
  • For depletion of cells using a single antibody, we recoomend the MagniSort™ Streptavidin Positive Selection Beads (cat. MSPB-6003) and protocol
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 Tests
Streptavidin
Biotin
Liquid
Streptavidin Conjugate (Fluorescent)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.