Learn More
Bohlender™ Magnetic Stirrer Head (G-MRK)
Ideal for reactor lids with a center ground joint. BOLA Magnetic Stirrer Heads are suitable for stirrer shafts made of glass or stainless steel with counterbores for a safe fixing into the rotor.
Brand:  Bohlender™ C502-16
Product Code. 10399239
  • 1800.00€ / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Description
  • Perfect combination of drive shaft with ball bearings, rotor and lower bearing made of PTFE/PEEK and a conductor made of borosilicate glass
  • Requires little space due to compact construction
  • No leakage or memory effects due to non-porous, welded rotor
  • Rotor holds the stirrer shaft by means of three stud screws which are fixed in the counterbores of the stirrer shaft
  • This provides optimum power transmission and a safe fixing
  • 6mm square can be fixed into the stirrer coupling or into the agitator
  • Powerful transmission
  • No grease required
  • All products which are exposed to the medium do not contain any metals
  • High speed of up to max. 1.500 rpm
  • High working temperatures up to +250°C are possible
  • Excellent chemical resistance
  • Safe to run dry
  • Long durability
  • Space-saving drive shaft O.D.'s
Specifications
Specifications
Magnetic Stirrer Head