missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate heptahydrate, meets analytical spec. of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance), Honeywell Fluka™
BIOPHARMA

Product Code. 15681250
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
6 x 2.5 kg

Meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Quantity 5 kg
Linear Formula MgSO4 · 7H2O
UN Number NONH for all modes of transport
Formula Weight 246.47g/mol
Percent Purity 99.5 to 100.5% (calc. to the dried substance)
Packaging Plastic Bottle
Vapor Pressure 0.1mmHg (20°C)
Density Vapor Density: <0.01 (vs air)
Chemical Name or Material Magnesium Sulfate Heptahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.