missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium sulfate heptahydrate, meets analytical spec. of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance), Honeywell Fluka™
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 100 g |
Linear Formula | MgSO4 · 7H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 246.47g/mol |
Percent Purity | 99.5 to 100.5% (calc. to the dried substance) |
Packaging | Plastic Bottle |
Vapor Pressure | 0.1mmHg (20°C) |
Density | Vapor Density: <0.01 (vs air) |
Chemical Name or Material | Magnesium Sulfate Heptahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'