missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, tested according to Ph.Eur, Honeywell™
BIOPHARMA

Product Code. 15686070
Click to view available options
Quantity:
500 g

Tested according to Ph.Eur., light

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 1309-48-4
Quantity 500 g
Molecular Formula MgO
Linear Formula MgO
MDL Number MFCD00011109
UN Number NONH for all modes of transport
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
SMILES O=[Mg]
IUPAC Name oxomagnesium
Molecular Weight (g/mol) 40.30
PubChem CID 14792
Formula Weight 40.3g/mol
Chemical Name or Material Magnesium oxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'