missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals

62.91€ - 203.01€

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10741471
View Documents
Thermo Scientific Acros
193430100
10 g Glass bottle
62.91€
10g
Please to purchase this item. Need a web account? Register with us today!
10249833
View Documents
Thermo Scientific Acros
193430500
50 g Glass bottle
203.01€
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

1309-48-4
40.30
CPLXHLVBOLITMK-UHFFFAOYSA-N
14792
O=[Mg]
MgO
MFCD00011109
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
oxomagnesium

Specifications

3600.0°C
1309-48-4
Powder
MgO
MFCD00011109
15, 5741
Solubility in water: 6.2 mg/l (20°C), reacts generates heat and mg(oh)2. Other solubilities: soluble in diluted acids, insoluble in alcohol
O=[Mg]
40.30
40.29
Trace Metal Basis
3.5800g/mL
2800.0°C
White
99.99%
MgO
01,633; 02,256; 07,221; 16,200
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
CPLXHLVBOLITMK-UHFFFAOYSA-N
oxomagnesium
14792
99.99%
3.58
Magnesium oxide, Extra Pure
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-171-9

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.