missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAD2L1 Antibody (1761), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43820
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
MAD2L1 Monoclonal specifically detects MAD2L1 in Human samples. It is validated for Western Blot.
Specifications
| MAD2L1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| MAD2L1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human MAD2L1. The exact sequence is proprietary. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 4085 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot | |
| 1761 | |
| Western Blot 1:100-1:3000 | |
| HSMAD2, MAD2 (mitotic arrest deficient, yeast, homolog)-like 1, MAD2 mitotic arrest deficient-like 1 (yeast), MAD2-like protein 1, MAD2mitotic arrest deficient, yeast, homolog-like 1, Mitotic arrest deficient 2-like protein 1, mitotic spindle assembly checkpoint protein MAD2A | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction