Learn More
Cytiva Macrosep™ Advance Centrifugal Devices with Supor Membrane
Product Code. 17144741
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Includes:
100/Pk.
24/Pk.
Unit Size:
Pack of 100
Pack of 24
Product Code. 17144741
Brand: Cytiva MAPM45C67
Only null left
Add to basket
Add to basket
Particulate removal for longer HPLC and UHPLC column life.
  • High spin speed and larger EFA reduces spin times
  • For samples from 3 to 20 mL
  • For research applications only
TRUSTED_SUSTAINABILITY
Specifications
Color Wildberry
Description EFA: 7.2 cm2
Material (Membrane) PES
Pore Size 0.45 μm
Sterility Non-sterile
Includes 24/Pk.
Volume (Metric) 5 to 20 mL (Sample)