Learn More
Thermo Scientific™ MAC 1753
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Quantity | 5 mg |
| Content And Storage | Ambient temperatures |
| For Use With (Application) | A non-peptide GnRH agent capable of inhibiting the effect of gonadotropin-releasing hormone |
| CAS | 685830-90-4 |
| Molecular Formula | C16H10Cl2N2O3 |
| Formula Weight | 349.17 |
| Synonym | 5-(3,5-Dichlorophenoxy)-N-4-pyridinyl-2-furancarboxamide; |
| Product Type | MAC 1753 |