Learn More
Brady™ M710 Label Printer with Wi-Fi and Bluetooth
More speed, less mess. It’s Brady™'s fastest, most advanced portable printer yet with a print speed of 76.2 mm/sec with Wi-Fi and Bluetooth.
Brand:  Brady™
 View more versions of this product
Product Code. 17895235
  • 1060.00€ / Each
Explore more special offers
Description
Description
  • Auto-cut labels and work all day on one charge
  • Use one device for 100's of applications, printing on the toughest materials around
  • Stay connected wherever your work happens with enhanced Bluetooth and Wi-Fi
  • Get lightning-fast print speeds, plus cut labels automatically
  • Prints 4,500 labels on a full battery charge
  • Enjoy more durability with a printer that withstands 1.2 m drops, blowing sand and dust
  • Get more work done using a keypad or software to print labels
  • Print what you need, how you need – text, barcodes, shapes and images using a keypad or software
  • Get shop-to-field flexibility with label saving, sharing and data imports
Specifications
Specifications
Printer
Color LCD Touchscreen
Plastic
4.6 in.
116.75 mm
13.59 in.
Monocolor
Printer, Hard Case, USB Cable, Quick Start Guide, Drivers, Li-ion Rechargeable Battery, AC Adapter, Users Guide, Brady Workstation Basic Design Software, Media Adapter, Roll of M7C-1000-595-WT and M7-R6000 Ribbon
200 MB (Stores Up to 100,000 Labels)
76.2 mm/sec.
Bluetooth, Wi-Fi, USB 2.0, USB-C
2.45 kg
Electrical Labeling, Facility Identification, General Identification, Healthcare and Hospital Labeling, Laboratory Labeling
AC Adaptor, Rechargeable Battery
QWERTZ
Blue
Brady Labels
Thermal Transfer
9.45 in.
240.03 mm
345.2 mm
53.77 x 71.28 mm
English (US), French, Spanish, Portuguese, German, Dutch, Norwegian, Finnish, Swedish, Italian, Danish, Bulgarian, Russian, Croatian, Hungarian, Japanese, Korean, Polish, Romanian, Slovak, Turkish, Estonian, Chinese (Traditional), Chinese (Simplified)
M710
300 dpi
5.4 lb.
FCC Part 15 Class A
2 Years
110/240 V