Learn More
Brady™ M710 Label Printer with BWS SFID Suite
More speed, less mess. It’s Brady™'s fastest, most advanced portable printer yet with a print speed of 76.2 mm/sec. Printer includes Brady™ WorkStation Product & Wire Identification Suite.
Brand:  Brady™ 317826
 View more versions of this product
Product Code. 17845255
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Auto-cut labels and work all day on one charge
  • Use one device for 100's of applications, printing on the toughest materials around
  • Get lightning-fast print speeds, plus cut labels automatically
  • Work longer – print 4,500 labels on a full battery charge
  • Enjoy more durability with a printer that withstands 1.2 m drops, blowing sand and dust
  • Print what you need, how you need – text, barcodes, shapes and images using a keypad or software
  • Get shop-to-field flexibility with label saving, sharing and data imports
  • Comes with a Brady™ WorkStation Safety & Facility Identification Suite, enabling you to immediately start designing professional labels
Specifications
Specifications
Printer
Color LCD Touchscreen
Plastic
4.6 in.
116.75 mm
13.59 in.
Monocolor
Printer, Hard Case, USB Cable, Quick Start Guide, Drivers, Li-ion Rechargeable Battery, AC Adapter, Users Guide, Brady WorkStation Safety and Facility ID Software Suite Download, Media Adapter, Roll of M7C-1000-595-WT and M7-R6000 Ribbon
200 MB (Stores Up to 100,000 Labels)
76.2 mm/sec.
USB 2.0, USB-C
2.45 kg
Facility Identification, Safety Identification
AC Adaptor, Rechargeable Battery
AZERTY
Blue
Brady Labels
Thermal Transfer
9.45 in.
240.03 mm
345.2 mm
53.77 x 71.28 mm
English (US), French, Spanish, Portuguese, German, Dutch, Norwegian, Finnish, Swedish, Italian, Danish, Bulgarian, Russian, Croatian, Hungarian, Japanese, Korean, Polish, Romanian, Slovak, Turkish, Estonian, Chinese (Traditional), Chinese (Simplified)
M710
300 dpi
5.4 lb.
FCC Part 15 Class A
2 Years
110/240 V