Learn More
Applied Biosystems™ M13 Sequencing Forward Primer, HPLC
M13 Sequencing Forward Primer, HPLC
Brand: Applied Biosystems™ A15637
This item is not returnable.
View return policy
Description
Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.Specifications
Single Tube, Dried-down | |
M13 (-21) Forward Sequencing Primer, 50 nmole, HPLC Purification, 900pmole Minimal Spec Yield, 5' TGTAAAACGACGGCCAGT Sequence, Delivered in a Single tube, Dried-down, Used for BigDye™ Cycle Sequencing Primers | |
5' TGTAAAACGACGGCCAGT | |
50 nmol | |
BigDye™ Cycle Sequencing Primers |
Mass Spectrometry | |
M13 (-21) Forward Sequencing Primer | |
M13 (-21) Forward Sequencing | |
HPLC | |
Solid |
For Research Use Only. Not for use in diagnostic procedures.