Learn More
Applied Biosystems™ M13 Human, Reverse Primer, HPLC
M13 Human, Reverse Primer, HPLC
Brand:  Applied Biosystems™ A15636
Product Code. 15899866
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
M13 Human Reverse HPLC Primer, 50 nmole, Solid, For PCR and Sanger Sequencing
Reverse
HPLC
M13 Human Reverse HPLC Primer
50 nmol
Solid