Learn More
Applied Biosystems™ M13 Human, Forward Primer, HPLC
M13 Human, Forward Primer, HPLC
Brand: Applied Biosystems™ A15635
This item is not returnable.
View return policy
Description
Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.Specifications
M13 Human Forward HPLC Primer, 50 nmole, For PCR and Sanger Sequencing | |
Forward | |
HPLC |
M13 Human Forward HPLC Primer | |
50 nmol |
For Research Use Only. Not for use in diagnostic procedures.