missing translation for 'onlineSavingsMsg'
Learn More

m-Anisic acid, 98%

Product Code. 10380544 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10380544

Brand: Thermo Scientific Acros 152475000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 586-38-9 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-38-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002499
InChI Key XHQZJYCNDZAGLW-UHFFFAOYSA-N
Synonym m-anisic acid, m-methoxybenzoic acid, benzoic acid, 3-methoxy, 3-anisic acid, meta-anisic acid, 3-methoxy-benzoic acid, unii-e2i36fh6qz, m-methylsalicylic acid, e2i36fh6qz, chembl22425
PubChem CID 11461
IUPAC Name 3-methoxybenzoic acid
SMILES COC1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 103.0°C to 108.0°C
Color Beige-Yellow to White
Boiling Point 170.0°C to 172.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula CH3OC6H4CO2H
Quantity 500 g
Beilstein 10, 137
Formula Weight 152.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material m-Anisic acid
Show More Show Less
compliance-icons
Product Identifier
  • m-Anisic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.