missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysing Matrix Y, Bulk

Lysing Matrix Y is composed of 0.5 mm diameter Yttria-Stabilized Zirconium Oxide beads and is used primarily for lysis of gram positive and gram negative bacteria, fungal tissue and spores, yeasts, molds, filamentous algaes and biofilms.

Brand:  MP Biomedicals™ 116540436

512.66 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15782282

  • 552.00€ / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Applications: Lysing Matrix Y is used primarily for lysis of gram positive and gram negative bacteria, fungal tissue and spores, yeasts, molds, filamentous algaes and biofilms.
  • The beads have a high density and an extremely high hardness making this matrix ideal for cracking of very tough cell walls while its smooth surface minimizes shearing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.5 mm Yttria-stabilized Zirconium Oxide Beads
Yeast, Fungi, Algea
Nucleic Acid Isolation and Purification, Homogenization
500 g
0.5 mm Yttria-Stabilized Grinding Beads, Y Matrix
FastPrep instruments or other bead beaters
Lysing Matrix Y
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.