missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LYN 1604 dihydrochloride
Potent UNC-51-like kinase 1 (ULK1) agonist
168.00€ - 625.00€
Specifications
Chemical Name or Material | 2-[Bis(2-methylpropyl)amino]-1-[4-[2-(2,4-dichlorophenyl)-2-(2-naphthalenylmethoxy)ethyl]-1-piperazinyl]ethanone dihydrochloride |
---|---|
CAS | 2310109-38-5 |
Target | ULK1 |
Molecular Formula | C33H43Cl2N3O2.2HCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18747706
|
Tocris Bioscience™
6617/10 |
10 mg |
168.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18748106
|
Tocris Bioscience™
6617/50 |
50 mg |
625.00€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LYN 1604 dihydrochloride is a potent UNC-51-like kinase 1 (ULK1) agonist (EC50 = 18.94 nM). In MDA-MB-231 cells, LYN 1604 induces upregulation of Beclin-1, LC3-I and LC3-II, as well as degradation of p62. LYN 1604 induces cell death (IC50 = 1.66 μM) via the ULK complex involving both autophagy and apoptosis pathway effectors (e.g. ATF3, RAD21, and caspase-3); this effect is reversible by the autophagy inhibitor 3-methyladenine (Cat. No. 3977). LYN 1604 inhibits tumor growth in a mouse xenograft model, decreases bone loss in a mouse model of osteoporosis and reduces the formation of TRAP-positive multinucleated cells.Specifications
2-[Bis(2-methylpropyl)amino]-1-[4-[2-(2,4-dichlorophenyl)-2-(2-naphthalenylmethoxy)ethyl]-1-piperazinyl]ethanone dihydrochloride | |
2310109-38-5 | |
ULK1 | |
C33H43Cl2N3O2.2HCl |
Safety and Handling
Recommended Storage : Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title