missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LY 255283, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective, competitive antagonist of BLT2 receptors (IC50 values are ∽ 1 and > 10 μM at human recombinant BLT2 and BLT1 receptors respectively). Inhibits LTB4-induced contraction of lung parenchyma (pA2 = 7.2), and reduces LTB4-mediated airway obstruction in guinea pigs following i.v. and oral administration. .
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 360.46 |
Percent Purity | >98% |
Chemical Name or Material | LY 255283 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'