Learn More
Welch™ LVS 300 Z Laboratory Vacuum System
Product Code. 10364385 Shop All Welch Ilmvac Products
Click to view available options
Depth (Metric):
310 mm
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 10364385
Brand: Welch™ 115041
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulatory: This product is under eco-contribution tax.
Specifications
Electrical Requirements 230 V 50/60 Hz
Depth (Metric) 310 mm
Model LVS 300 Z
Frequency 50/60 Hz
Overall Height (Metric) 395 mm
Overall Width (Metric) 360 mm
Pump Type Laboratory Vacuum System
Maximum Pressure <8 mbar
Voltage 230 V