Learn More
WTW™ LR 925/01 2 Electrode Conductivity Cell
The conductivity measuring cells of the highest quality for decades. These are the basis for the new IDS conductivity measuring cells. The advantages of the IDS technology include the automatic transfer of the cell constant and therefore the exclusion of possible erroneous input.
Brand: WTW™ 301720
837.90 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.
Specifications
2-electrodes, Graphite | |
0°C to 100°C | |
120 mm |
1.5 m | |
12 mm |