Learn More
IKA™ LR 2000.10 Anchor Stirrer
Product Code. 10672372 Shop All IKA Products
Click to view available options
Product Type:
Anchor Stirrer
Unit Size:
Each
Product Code. 10672372
Brand: IKA™ 0002508400
Only null left
Add to basket
Add to basket
Use this vessel with IKA systems for the optimization and reproduction of various chemical reactions, mixing and homogenization processes. IKA™ LR 2000.10 Anchor Stirrer features a PEEK scraper and can be used with all laboratory reactors.
  • With PEEK scraper
  • For all laboratory reactors
  • Compatible with IKA LR-2.ST and LR 2000 V systems
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) For all Laboratory Reactors
Product Type Anchor Stirrer