Learn More
Fluid Metering™ Low Flow Isolation Kit
Brand:  Fluid Metering™ R479
Encompass
Product Code. 11974138
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Specifications
50 mL/min.
Low flow (below 50 mL/min.) applications where minimum system dead volume and maximum chemical inertness is required
Stainless Steel Pump Heads (Except San)
Four ferrules, two adapters and assembly/removal tools