Learn More
Lovibond™ Glass Filter Set for SpectroDirect & XD7000 Spectrophotometer
Complete Glass Filter Set for testing the photometric accuracy and the wavelength accuracy of the spectrophotometer
Brand:  Lovibond™ 711160
Product Code. 11715438
  • 1780.00€ / Each
Explore more special offers
Specifications
Secondary Standard Set
SP600 spectrophotometer
4 Glass filters for wavelength confirmation
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lovibond™ Glass Filter Set for SpectroDirect & XD7000 Spectrophotometer > Glass Filter Set SpectroDirect abd XD7000 Spectrophotometer