missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Ammonia Reagent Set, for Tube Testing, High Range

Premeasured unit dose reagent for water quality testing

Brand:  Lovibond™ 535650


Product Code. 13151086

  • 128.00€ / Pack of 50
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket

Includes: Ammonia Cyanurate Reagent (5mL sample, 100 pack), Ammonia Salicylate Reagent (5mL , 100 pack), HR AM Dilution Tube (50 ct.)
This item is not returnable. View return policy

Description

Description

Premeasured unit dose reagent for water quality testing

  • Offers same high quality standards as tablet reagents
  • Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open package
  • Easily create “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in corners

Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing

Specifications

Specifications

Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
5mL ammonia cyanurate reagent (100 pack), 5mL ammonia salicylate reagent (100 pack), 50 ct. HR AM dilution tube
0 to 50mg/L
High Range Ammonia Reagent Set
Hach™ and other manufacturers test kits, colorimeters and spectrophotometers
25
Salicylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Vario AM VIAL Test Reagent SET HR

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates
Promotions

Promotions