missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Powder Packs™ for Free Chlorine

Product Code. 11789029
Click to view available options
Quantity:
100 Pack
1000 Pack
Range:
0 to 2 mg/L
0 to 3.5 mg/L
0 to 5 mg/L
Volume (English):
0.16 oz.
0.33 oz.
0.84 oz.
Volume (Metric):
5 mL
10 mL
25 mL
Unit Size:
Pack of 100
Pack of 1000
This item is not returnable. View return policy

Product Code. 11789029

Brand: Lovibond™ 530103

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Premeasured unit dose DPD reagents for water quality testing

  • For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
  • Also suitable for use in Hach™ instruments and test kits
  • Easy-to-open, sealed foil packets eliminate risk of cross-contamination
  • Available for 5, 10, and 25mL sample sizes
  • Extended shelf life of up to 5 years

Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing

Compliance

US EPA-accepted DPD method

Specifications

Certifications/Compliance US EPA
Volume (Metric) 10 mL
Format Powder
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer
Quantity 1000 Pack
Range 0 to 2 mg/L
Type DPD Free Chlorine Reagent
Volume (English) 0.33 oz.
Hazard Statement
  • EUH210-Safety data sheet available on request.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.