Learn More
Lovibond™ 10 mm Path Length Glass Cells
A precision fused cuvette made from optical glass to the highest standards.
Brand: Lovibond™ 606810
This item is not returnable.
View return policy
Description
- 10mm path length
- For measuring the color of varnishes, drying oils, resins, nitro-cellulose, lacquers, fatty acids and similar transparent colored liquids
- Complies with ASTM D-1544 and AOCS Method Td 1a
Specifications
Glass Cells | |
Comparator 2000+, comparator 3000 with non-heated Samples |
Product Content Correction
Product Title
Lovibond™ 10 mm Path Length Glass Cells > Glass Cells, 10mm Path Length
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur