missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ 293 Cell Serum-Free Media
Completely balanced to support vaccine and viral vector production of high-density human embryonic kidney (HEK) 293 cell culture environments
166.00€
Specifications
Content And Storage | 2° to 8°C |
---|---|
Without Additives | No Glutamine, No Phenol Red |
Format | Bottle |
Quantity | 1 L |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
11675250
|
Lonza™
BE12-764Q |
Pro293A-CDM, for adherent cells | N/A | ||||||
Description
Very low levels of recombinant proteins facilitate downstream purification and provide regulatory compliance
- Chemically-defined media optimized for specific cell types
Specifications
2° to 8°C | |
No Glutamine, No Phenol Red | |
Bottle | |
1 L |