Learn More
Testo™ ComSoft Basic Software
Product Code. 10241790
Click to view available options
For Use With (Equipment):
ComSoft Basic
Unit Size:
Each
Product Code. 10241790
Brand: Testo™ 0572 0580
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Use this software with the instrument series Testo 174, Testo 175 and Testo 176. Testo™ ComSoft Basic Software features graphic and tabular measurement value presentation as well as export function.
  • Software for programming and readout of Testo data loggers
  • Free download of the ComSoft Basic (with registration) at www.testo.com
  • Offers all basic functions of a logger software
  • Runs on Windows XP ServicePack 3 (SP3), Windows Vista, Windows 7 and more
  • Intuitive user interface guides the user through the individual processes step by step
  • Graphic measurement value curve for clear presentation of the measurement values
  • Logger configuration: start and stop criterion, measurement interval, storage rate and limit value adjustment
  • Convenient export functions e.g. for the further processing of the data in Microsoft Excel, or the generation of a PDF which can be made available to other users
Specifications
Type Datalogger Software
For Use With (Application) Programming and readout of Testo data loggers, graphic and tabular measurement value presentation as well as export function
For Use With (Equipment) ComSoft Basic