Learn More
Invitrogen™ LiveBLAzer™ FRET-B/G Loading Kit with CCF2-AM
LiveBLAzer™ FRET-B/G Loading Kit with CCF2-AM
Brand:  Invitrogen™ K1025
Product Code. 11584906
  • 30590.00€ / 5mg
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
LiveBLAzer™ FRET-B/G Loading Kit with CCF2-AM
Background:
GeneBLAzer™ cell-based assays utilize the membrane-permeant ester forms (CCF2-AM and CCF4-AM) of the negatively charged fluorescent beta-lactamase substrates, CCF2 and CCF4. These lipophilic esters readily enter the cell, where cleavage by endogeneous cytoplasmic esterases rapidly converts them into their negatively charged forms, thereby trapping them in the cytosol.
Detection of GeneBLAzer™ assays is FRET-based. Each substrate is labeled with two fluorophores that form an efficient FRET pair. In the absence of beta-lactamase activity, exciting the coumarin at 409 nm in the intact CCF2 molecule results in FRET to the fluorescein, which emits a green fluorescence signal at 518 nm (Figure 1). In the presence of beta-lactamase activity, however, cleavage of CCF2 spatially separates the two dyes and disrupts FRET, so that exciting the coumarin at 409 nm now produces a blue fluorescence signal at 447 nm. This blue signal can be readily observed under a microscope and can also be detected as an increase in the blue channel readout on fluorescent microplate readers.
The CCF2-AM and CCF4-AM substrates are essential assay components for the GeneBLAzer™ platform. These substrates are fully compatible with flow cytometry, speeding the time to clone selection. Ratiometric analysis of the blue and green signals reduces well-to-well variation due to differences in cell numbers and substrate loading, leading to high Z´-factor values and low coefficients of variation (CVs).
CCF2-AM and CCF4-AM differ by two carbons in the bridge linking the coumarin moiety to the lactam ring. Both are in the membrane-permeable, esterified forms, and can be used for assays in intact cells. CCF4-AM has better solubility properties (soluble for >24 hours) than CCF2-AM and is thus best suited for screening applications. In addition, CCF4-AM has slightly better FRET and thus slightly lower background than CCF2-AM.
CCF2-FA is essentially the CCF2 substrate without the esters found in the AM version. CCF2-FA is de-esterified and used in cell lysate applications, bypassing loading across the cell membrane and de-esterification steps. Cell lysates are the preferred method for applications using cells that contain a cell wall. CCF2-FA can also be used as a control to acquire the excitation and emission spectra for CCF2-AM and CCF4-AM.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
–20°C
Fluorescent
GeneBLAzer™ beta-lactamase substrates are supplied as loading kits (Table 1) and are available in bulk sizes. Loading kits include CCF2-AM or CCF4-AM, DMSO (for Solution A), Solution B and Solution C. CCF2-AM and CCF4-AM are provided dry and should be stored at –20°C, desiccated and protected from light. DMSO (for Solution A), Solution B and Solution C should be stored at 22-25°C, protected from direct light. Slightly cooler temperatures may cause a white precipitate to form in Solution B
5mg
LiveBLAzer™ FRET-B/G Loading Kit with CCF2-AM, 5mg, CCF2-AM Substrate, Chemical Substrate Properties, Fluorescent Detection Method, Cell-Permeant Permeability, –20°C Storage
Cellular imaging, pharma and biopharma, protein assays and analysis, Protein biology, cell analysis, cellular imaging, cell-based reporter assays, cell-based and beta-lactamase assays, drug discovery and development, target and lead identification and validation, kinase biology, kinase pathway analysis, g-protein coupled receptor biology, cell-based GPCR reporter assays, nuclear receptor biology, cell-based nuclear receptor reporter assays, pathway biology, cellular pathway analysis assays, other cellular pathway assay products, enzyme reporter assays, protein assays and analysis, beta-lactamase activity assays
LiveBLAzer™ FRET-B/G Loading Kit with CCF2-AM
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.