missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Litmus, Indicator, pH 5-8, for titration, Honeywell Fluka™
Indicator, pH 5-8, for titration
154.92€ - 519.10€
Specifications
Chemical Name or Material | Litmus |
---|---|
Name Note | Indicator, pH 5-8, for titration |
CAS | 1393-92-6, 64-17-5 |
CAS Max % | ≤4.0000%, ≤100.0000% |
CAS Min % | ≥96.0000% |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Litmus | |
1393-92-6, 64-17-5 | |
≥96.0000% | |
I2 | |
Analytical Reagent |
Indicator, pH 5-8, for titration | |
≤4.0000%, ≤100.0000% | |
I2 | |
MFCD00011355 | |
Glass Bottle |
Safety and Handling
missing translation for 'shelfLife' : 1620 days from date of manufacture
Recommended Storage : Room Temp