Learn More
Lithium diisopropylamide mono(tetrahydrofuran), 1.5M solution in cyclohexane, AcroSeal™, Thermo Scientific™
54.30€ - 197.20€
Chemical Identifiers
CAS | 123333-84-6, 110-82-7 |
---|---|
Molecular Formula | C10H22LiNO |
Molecular Weight (g/mol) | 179.23 |
InChI Key | YTJXGDYAEOTOCG-UHFFFAOYSA-N |
Synonym | lithium diisopropylamide mono tetrahydrofuran, lithium diisopropylamide mono tetrahydrofuran complex solution, lithium;di propan-2-yl azanide;oxolane, acmc-20ehmk, lda thf, lda.thf, lithiumdiisopropylamide thf, lithium diisopropylamide thf, lithiumdiisopropylamide tetrahydrofuran, lithium diisopropylamide tetrahydorfuran |
PubChem CID | 10997623 |
IUPAC Name | lithium;di(propan-2-yl)azanide;oxolane |
SMILES | [Li+].CC(C)[N-]C(C)C.C1CCOC1 |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
123333-84-6, 110-82-7 | |
179.23 | |
lithium diisopropylamide mono tetrahydrofuran, lithium diisopropylamide mono tetrahydrofuran complex solution, lithium;di propan-2-yl azanide;oxolane, acmc-20ehmk, lda thf, lda.thf, lithiumdiisopropylamide thf, lithium diisopropylamide thf, lithiumdiisopropylamide tetrahydrofuran, lithium diisopropylamide tetrahydorfuran | |
lithium;di(propan-2-yl)azanide;oxolane |
C10H22LiNO | |
YTJXGDYAEOTOCG-UHFFFAOYSA-N | |
10997623 | |
[Li+].CC(C)[N-]C(C)C.C1CCOC1 |
Specifications
123333-84-6, 110-82-7 | |
Liquid | |
−18°C | |
YTJXGDYAEOTOCG-UHFFFAOYSA-N | |
lithium;di(propan-2-yl)azanide;oxolane | |
10997623 | |
1.43 to 1.65M | |
0.8160g/mL |
Brown to Orange-Yellow | |
C10H22LiNO | |
lithium diisopropylamide mono tetrahydrofuran, lithium diisopropylamide mono tetrahydrofuran complex solution, lithium;di propan-2-yl azanide;oxolane, acmc-20ehmk, lda thf, lda.thf, lithiumdiisopropylamide thf, lithium diisopropylamide thf, lithiumdiisopropylamide tetrahydrofuran, lithium diisopropylamide tetrahydorfuran | |
[Li+].CC(C)[N-]C(C)C.C1CCOC1 | |
179.23 | |
179.23 | |
0.816 | |
Lithium diisopropylamide mono(tetrahydrofuran) |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Lithium diisopropylamide mono(tetrahydrofuran), 1.5M solution in cyclohexane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.