missing translation for 'onlineSavingsMsg'
Learn More

Liquiritigenin, Thermo Scientific™

Product Code. p-7227397 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Product Code. 17730679

Brand: Thermo Scientific Acros 469812500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Specifications

Alpha Vector LIQUIRITIGENIN
Chemical Name or Material Liquiritigenin
CAS 578-86-9
Color Beige to White
Boiling Point 530°C (760 mmHg)
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C15 H12 O4
MDL Number 00287289
Quantity 250 mg
Synonym 4 ',7-Dihydroxyflavanone
Molecular Weight (g/mol) 256.26
Physical Form Crystals or Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.