Learn More
Novus Biologicals™ Lightning-Link (R) R-PE Antibody Labeling Kit
Applications: Flow Cytometry
492.00€ - 3205.00€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18785583
|
Novus Biologicals™
703-0003 |
5 x 1 mg Reaction |
3205.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18749963
|
Novus Biologicals™
703-0004 |
1 x 5 mg Reaction |
3050.00€
3mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18765403
|
Novus Biologicals™
703-0005 |
1 x 100 μg Reaction |
492.00€
60µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18322558
|
Novus Biologicals™
703-0015 |
1 mg Reaction |
1090.00€
600µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18715153
|
Novus Biologicals™
703-0030 |
3 x 0.01 mg Reaction |
494.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18747973
|
Novus Biologicals™
703-0010 |
3 x 60 μg Reaction |
745.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
R-Phycoerythrin (R-PE) is a fluorescent protein from the phycobiliprotein family, and is isolated from red algae. The absorbance spectrum of R-PE covers a broad range of excitation wavelengths, which provides an advantage for multi-color immunofluorescent staining or cell sorting. R-PE is one of the most intensely fluorescent phycobiliprotein having orange fluorescence. It is significantly brighter and more photostable than conventional organic fluorophores and it has a high quantum yield.Specifications
PE |
Product Title
Novus Biologicals™ Lightning-Link (R) R-PE Antibody Labeling Kit