Learn More
Invitrogen™ Novex™ IL-10 Human Ultrasensitive Magnetic Bead Kit
Designed for quantification of low levels of human IL-10 in serum, plasma and tissue culture supernatant samples
Brand:  Invitrogen™ LHC0103M
Product Code. 15314584
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Includes: Antibody coated capture beads, standard, detector antibody and lot-specific technical data sheet