Learn More
Invitrogen™ Novex™ IL-1β Human Ultrasensitive Magnetic Bead Kit
Designed for quantification of low levels of human IL-2β in serum, plasma and tissue culture supernatant samples. Invitrogen™ Novex™ IL-1β Human Ultrasensitive Magnetic Bead Kit is an ideal solution for assessing multiple protein biomarkers in a single sample.
Brand:  Invitrogen™ LHC0013M
Product Code. 15364574
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Includes: Antibody coated capture beads, standard, detector antibody and lot-specific technical data sheet
Description
Description
This immunoassay uses Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins. This assay is subject to the same stringent and robust development and manufacturing as are the extensively validated Platinum and Instant ELISA kits. Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma.